ENA018106194 Enamine

5-{[3-(trifluoromethyl)phenyl]methyl}-1,3-thiazol-2-amine

  • MDL number MFCD00709518
  • Cas Number : 300819-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65-66
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02545

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02545
Availability details for
Quantity
  • Availability
  • Add to Cart