ENA018106236 Enamine

4-[(2-methoxyphenoxy)methyl]-1,3-thiazol-2-amine

  • MDL number MFCD06660611
  • Cas Number : 879151-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-02606

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02606
Availability details for
Quantity
  • Availability
  • Add to Cart