ENA018106237 Enamine

4-[(4-methylphenoxy)methyl]-1,3-thiazol-2-amine

  • MDL number MFCD06660612
  • Cas Number : 885457-99-8
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02607

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02607
Availability details for
Quantity
  • Availability
  • Add to Cart