ENA018106238 Enamine

4-[(2-methylphenoxy)methyl]-1,3-thiazol-2-amine

  • MDL number MFCD06660613
  • Cas Number : 879145-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95...97
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02608

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02608
Availability details for
Quantity
  • Availability
  • Add to Cart