ENA018106239 Enamine

4-[(4-bromophenoxy)methyl]-1,3-thiazol-2-amine

  • MDL number MFCD06660614
  • Cas Number : 885458-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-02609

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02609
Availability details for
Quantity
  • Availability
  • Add to Cart