ENA018106240 Enamine

4-[(2,4-dichlorophenoxy)methyl]-1,3-thiazol-2-amine

  • MDL number MFCD03719765
  • Cas Number : 612802-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02610

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02610
Availability details for
Quantity
  • Availability
  • Add to Cart