ENA018106243 Enamine

ethyl 3-amino-4-(1H-imidazol-1-yl)benzoate

  • MDL number MFCD06660617
  • Cas Number : 875160-11-5
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91-92
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-02614

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02614
Availability details for
Quantity
  • Availability
  • Add to Cart