ENA018106245 Enamine

3-methoxy-N-({5H,6H,7H,8H,9H-[1,2,4]triazolo[4,3-a]azepin-3-yl}methyl)aniline

  • MDL number MFCD03655896
  • Cas Number : 750614-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-179
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02616

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02616
Availability details for
Quantity
  • Availability
  • Add to Cart