ENA018106254 Enamine

4-(4-ethoxyphenyl)-5-methyl-1,3-thiazol-2-amine

  • MDL number MFCD00994479
  • Cas Number : 443922-53-0
  • Molecular Weight : 234.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-02633

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02633
Availability details for
Quantity
  • Availability
  • Add to Cart