ENA018106256 Enamine

4-amino-N-(pyrrolidin-2-ylidene)benzene-1-sulfonamide

  • Cas Number : 102873-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02635

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02635
Availability details for
Quantity
  • Availability
  • Add to Cart