ENA018106279 Enamine

5-Amino-4-(1H-benzoimidazol-2-yl)-1-(2-morpholin-4-yl-ethyl)-1,2-dihydro-pyrrol-3-one

  • MDL number MFCD06641873
  • Cas Number : 459213-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02666

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02666
Availability details for
Quantity
  • Availability
  • Add to Cart