ENA018106282 Enamine

5-Amino-4-(1H-benzoimidazol-2-yl)-1-(4-chloro-phenyl)-1,2-dihydro-pyrrol-3-one

  • MDL number MFCD06660625
  • Cas Number : 459137-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 265...267
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02669

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02669
Availability details for
Quantity
  • Availability
  • Add to Cart