ENA018106292 Enamine

4,6-Diamino-1-(4-fluoro-phenyl)-2-oxo-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine-5-carbonitrile

  • MDL number MFCD06660631
  • Cas Number : 878466-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02693

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02693
Availability details for
Quantity
  • Availability
  • Add to Cart