ENA018106305 Enamine

N-(5-amino-3-oxo-4-phenyl-2,3-dihydro-1H-pyrrol-1-yl)benzamide

  • MDL number MFCD06660635
  • Cas Number : 885458-25-3
  • Molecular Weight : 293.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-02706

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02706
Availability details for
Quantity
  • Availability
  • Add to Cart