ENA018106315 Enamine

Thiophene-2-carboxylic acid [5-amino-4-(1H-benzoimidazol-2-yl)-3-oxo-2,3-dihydro-pyrrol-1-yl]-amide

  • MDL number MFCD06660641
  • Cas Number : 885458-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 270...272
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02716

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02716
Availability details for
Quantity
  • Availability
  • Add to Cart