ENA018106323 Enamine

3,4-diamino-N,N-diethylbenzene-1-sulfonamide

  • MDL number MFCD03150675
  • Cas Number : 49804-28-6
  • Molecular Weight : 243.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124-125
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-02743

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-02743
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart