ENA018106332 Enamine

N-(2-methoxyphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine

  • MDL number MFCD03069531
  • Cas Number : 27779-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-84
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-02752

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02752
Availability details for
Quantity
  • Availability
  • Add to Cart