ENA018106337 Enamine

(4,5-Dihydro-thiazol-2-yl)-(4-ethoxy-phenyl)-amine

  • MDL number MFCD03152761
  • Cas Number : 145621-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02757

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02757
Availability details for
Quantity
  • Availability
  • Add to Cart