ENA018106341 Enamine

N-(2-methylpropyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD03152764
  • Cas Number : 24622-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-02761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02761
Availability details for
Quantity
  • Availability
  • Add to Cart