ENA018106344 Enamine

6-amino-1-benzyl-5-(benzylamino)-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD02368562
  • Cas Number : 379725-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02764

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02764
Availability details for
Quantity
  • Availability
  • Add to Cart