ENA018106347 Enamine

2-(2-amino-1,3-thiazol-4-yl)-5-methoxyphenol

  • MDL number MFCD03475947
  • Cas Number : 199602-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02767

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02767
Availability details for
Quantity
  • Availability
  • Add to Cart