ENA018106358 Enamine

5-amino-N-(3-bromo-4-ethoxyphenyl)-2-methoxybenzene-1-sulfonamide

  • MDL number MFCD03478377
  • Cas Number : 438031-69-7
  • Molecular Weight : 401.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-02779

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02779
Availability details for
Quantity
  • Availability
  • Add to Cart