ENA018106367 Enamine

bis(4-methylphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD03655002
  • Cas Number : 102026-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153-154
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02788

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02788
Availability details for
Quantity
  • Availability
  • Add to Cart