ENA018106369 Enamine

N-butyl-1,3-benzothiazol-2-amine

  • MDL number MFCD00613459
  • Cas Number : 24622-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-02790

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02790
Availability details for
Quantity
  • Availability
  • Add to Cart