ENA018106374 Enamine

N-(4-methylphenyl)-4H-3,1-benzothiazin-2-amine

  • MDL number MFCD03478333
  • Cas Number : 109768-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-02795

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02795
Availability details for
Quantity
  • Availability
  • Add to Cart