ENA018106380 Enamine

N-(4-methoxyphenyl)-4,4-dimethyl-4,5-dihydro-1,3-thiazol-2-amine

  • MDL number MFCD03476333
  • Cas Number : 380436-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-02801

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02801
Availability details for
Quantity
  • Availability
  • Add to Cart