ENA018106388 Enamine

N-(3-methoxyphenyl)-4,4-dimethyl-4,5-dihydro-1,3-thiazol-2-amine

  • MDL number MFCD03476344
  • Cas Number : 380437-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-106
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-02809

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02809
Availability details for
Quantity
  • Availability
  • Add to Cart