ENA018106390 Enamine

N-(3,4-dichlorophenyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD03478324
  • Cas Number : 438029-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161-162
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-02811

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02811
Availability details for
Quantity
  • Availability
  • Add to Cart