ENA018106396 Enamine

5-ethyl-N-(3-methoxyphenyl)-4,5-dihydro-1,3-thiazol-2-amine

  • MDL number MFCD03476329
  • Cas Number : 380436-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02817

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02817
Availability details for
Quantity
  • Availability
  • Add to Cart