ENA018106409 Enamine

4-methyl-N-[2-(propan-2-yl)phenyl]-4,5-dihydro-1,3-thiazol-2-amine

  • MDL number MFCD03479188
  • Cas Number : 519150-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-02830

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02830
Availability details for
Quantity
  • Availability
  • Add to Cart