ENA018106412 Enamine

N-(2-ethoxyphenyl)-4,4-dimethyl-4,5-dihydro-1,3-thiazol-2-amine

  • MDL number MFCD03479180
  • Cas Number : 519150-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02833

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02833
Availability details for
Quantity
  • Availability
  • Add to Cart