ENA018106413 Enamine

N-(2,4-dimethylphenyl)-4-methyl-4,5-dihydro-1,3-thiazol-2-amine

  • MDL number MFCD03479178
  • Cas Number : 519150-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-02834

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02834
Availability details for
Quantity
  • Availability
  • Add to Cart