ENA018106416 Enamine

6-amino-1-benzyl-5-(butylamino)-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD03150845
  • Cas Number : 380433-20-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-02837

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02837
Availability details for
Quantity
  • Availability
  • Add to Cart