ENA018106425 Enamine

ethyl 2-amino-6-ethyl-4H,5H,6H,7H-thieno[2,3-c]pyridine-3-carboxylate

  • MDL number MFCD01556818
  • Cas Number : 281656-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75-76
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02846
Availability details for
Quantity
  • Availability
  • Add to Cart