ENA018106442 Enamine

6-Amino-1-benzyl-5-propylamino-1H-pyrimidine-2,4-dione

  • MDL number MFCD02167430
  • Cas Number : 343346-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210...212
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02863

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02863
Availability details for
Quantity
  • Availability
  • Add to Cart