ENA018106453 Enamine

N-cyclohexyl-5,6-dihydro-4H-1,3-thiazin-2-amine

  • MDL number MFCD03483909
  • Cas Number : 99977-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-02874

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02874
Availability details for
Quantity
  • Availability
  • Add to Cart