ENA018106455 Enamine

N,N-Dimethyl-3-(5-methyl-4,5-dihydro-thiazol-2-ylamino)-benzenesulfonamide

  • MDL number MFCD03483907
  • Cas Number : 522606-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111...113
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02876

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02876
Availability details for
Quantity
  • Availability
  • Add to Cart