ENA018106460 Enamine

6-amino-1-benzyl-5-{[(furan-2-yl)methyl]amino}-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD03965289
  • Cas Number : 726165-23-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02881

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02881
Availability details for
Quantity
  • Availability
  • Add to Cart