ENA018106464 Enamine

5-(benzylsulfanyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD00184237
  • Cas Number : 25660-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-111
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-02885

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02885
Availability details for
Quantity
  • Availability
  • Add to Cart