ENA018106466 Enamine

6-amino-1-benzyl-5-(cyclopentylamino)-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD03965300
  • Cas Number : 554404-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02887
Availability details for
Quantity
  • Availability
  • Add to Cart