ENA018106469 Enamine

6-Amino-1-benzyl-5-methylamino-1H-pyrimidine-2,4-dione

  • MDL number MFCD00205030
  • Cas Number : 72816-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219...221
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02890

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02890
Availability details for
Quantity
  • Availability
  • Add to Cart