ENA018106479 Enamine

(4,5-Dihydro-thiazol-2-yl)-(1-phenyl-ethyl)-amine

  • MDL number MFCD03479986
  • Cas Number : 143543-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02900

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02900
Availability details for
Quantity
  • Availability
  • Add to Cart