ENA018106483 Enamine

N-cyclohexyl-4-methylpyridin-2-amine

  • MDL number MFCD03651793
  • Cas Number : 522628-76-8
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49-50
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02904

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02904
Availability details for
Quantity
  • Availability
  • Add to Cart