ENA018106485 Enamine

bis(4-methoxyphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD02641119
  • Cas Number : 24827-38-1
  • Molecular Weight : 312.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-02906

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02906
Availability details for
Quantity
  • Availability
  • Add to Cart