ENA018106486 Enamine

4-methyl-N-phenyl-1,3-thiazol-2-amine

  • MDL number MFCD00226635
  • Cas Number : 90916-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74-75
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-02907

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02907
Availability details for
Quantity
  • Availability
  • Add to Cart