ENA018106490 Enamine

2-[(5-amino-1,3,4-thiadiazol-2-yl)sulfanyl]-N-(1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)acetamide

  • MDL number MFCD00374406
  • Cas Number : 329921-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-126
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02911

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02911
Availability details for
Quantity
  • Availability
  • Add to Cart