ENA018106497 Enamine

5-o-Tolyl-2H-tetrazole

  • MDL number MFCD02091280
  • Cas Number : 51449-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153...155
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02918

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-02918
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart