ENA018106502 Enamine

6-(piperazin-1-yl)-2,3,4,5-tetrahydro-1,2,4-triazine-3,5-dione

  • MDL number MFCD02293877
  • Cas Number : 568551-31-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-151
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-02923

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02923
Availability details for
Quantity
  • Availability
  • Add to Cart