ENA018106505 Enamine

2-(3-methoxyphenoxy)-5-(trifluoromethyl)aniline

  • MDL number MFCD03966849
  • Cas Number : 728907-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-02926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02926
Availability details for
Quantity
  • Availability
  • Add to Cart