ENA018106506 Enamine

2-(4-bromophenoxy)-5-(trifluoromethyl)aniline

  • MDL number MFCD03966882
  • Cas Number : 565193-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02927

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02927
Availability details for
Quantity
  • Availability
  • Add to Cart